A12482.0E
A12482.22
A12482.36
A12482.0EEA
392
EUR
InStock
A12482.0E
A12482.22
A12482.36
Cyanacetamid ≥99%
Cyanacetamid
Purity: 99%
CAS No.: 107-91-5
Molecular Formula: NCCH2CONH2
Formula Weight: 84.08
Flash Point: 215°(419°F)
Melting Point: 118-122°
Beilstein Reference: 878221
Merck Reference: 14,2688
MDL No.: MFCD00008024