A12505.0B
A12505.30
A12505.0BEA
108
EUR
InStock
A12505.0B
A12505.30
L(+)-Glutaminsäure Hydrochlorid ≥99%
L(+)-Glutaminsäure Hydrochlorid
Purity: 99%
CAS No.: 138-15-8
Molecular Formula: C5H9NO4.HCl
Formula Weight: 183.59
Melting Point: 203° dec.
Beilstein Reference: 3565569
Merck Reference: 14,4469
Optical Rotation: +23° (c=6 in water)
MDL No.: MFCD00012619