A12530.09
A12530.18
A12530.30
A12530.09EA
42.8
EUR
InStock
A12530.09
A12530.18
A12530.30
(4-Chlorphenyl)acetylchlorid ≥98%
(4-Chlorphenyl)acetylchlorid
Purity: 98%
CAS No.: 25026-34-0
Molecular Formula: C8H6Cl2O
Formula Weight: 189.04
Flash Point: 110°(230°F)
Refractive Index: 1.5510
Boiling Point: 112-114°/11mm
Beilstein Reference: 637775
UN No.: 3265
MDL No.: MFCD00037111