A12538.14
A12538.22
A12538.14EA
57.1
EUR
InStock
A12538.14
A12538.22
3-Nitro-p-toluidin ≥98%
3-Nitro-p-toluidin
Purity: 98%
CAS No.: 119-32-4
Molecular Formula: C7H8N2O2
Formula Weight: 152.15
Flash Point: 175°(347°F)
Odour: Characteristic
Beilstein Reference: 638984
UN No.: 2660
MDL No.: MFCD00007910