A12575.22
A12575.36
A12575.A3
A12575.22EA
39
EUR
InStock
A12575.22
A12575.36
A12575.A3
1-Methylimidazol ≥99%
1-Methylimidazol
Purity: 99%
CAS No.: 616-47-7
Molecular Formula: C4H6N2
Formula Weight: 82.11
Flash Point: 92°(197°F)
Refractive Index: 1.4970
Melting Point: -6°
Boiling Point: 195-197°
Beilstein Reference: 105197
UN No.: 3267
MDL No.: MFCD00005292