A12630.06
A12630.14
A12630.18
A12630.30
A12630.06EA
27.6
EUR
InStock
A12630.06
A12630.14
A12630.18
A12630.30
3-Methoxybenzylchlorid ≥98%
3-Methoxybenzylchlorid
Purity: 98+%
CAS No.: 824-98-6
Molecular Formula: C8H9ClO
Formula Weight: 156.61
Storage Temperature: 2°C to 8°C
Flash Point: 101°(213°F)
Refractive Index: 1.5440
Boiling Point: 122-123°/12mm
Beilstein Reference: 636684
UN No.: 1760
MDL No.: MFCD00000907