A12684.18
A12684.0B
A12684.30
A12684.18EA
31.5
EUR
InStock
A12684.18
A12684.0B
A12684.30
(1S)-(-)-Borneol ≥97%
(1S)-(-)-Borneol
Purity: 97+%
CAS No.: 464-45-9
Molecular Formula: C10H18O
Formula Weight: 154.25
Flash Point: 65°(149°F)
Physical Form: Cryst solid
Melting Point: 208-211°
Boiling Point: 210°
Beilstein Reference: 3587558
Merck Reference: 14,1338
Optical Rotation: -36° (c=5 in ethanol)
UN No.: 1312
MDL No.: MFCD00003759