A12686.06
A12686.14
A12686.06EA
37.7
EUR
InStock
A12686.06
A12686.14
2,3,5,6-Tetrafluorphenol ≥97%
2,3,5,6-Tetrafluorphenol
Purity: 97%
CAS No.: 769-39-1
Molecular Formula: C6H2F4O
Formula Weight: 166.07
Flash Point: 79°(174°F)
Odour: Phenol-like
Boiling Point: 141-143°
Beilstein Reference: 1911548
UN No.: 3261
MDL No.: MFCD00002157