A12688.14
A12688.22
A12688.36
A12688.14EA
41.9
EUR
InStock
A12688.14
A12688.22
A12688.36
2-Acetylpyridin ≥98%
2-Acetylpyridin
Purity: 98%
CAS No.: 1122-62-9
Molecular Formula: C7H7NO
Formula Weight: 121.14
Flash Point: 76°(169°F)
Refractive Index: 1.5220
Odour: Unpleasant
Melting Point: 8-10°
Boiling Point: 189-190°
Beilstein Reference: 107759
MDL No.: MFCD00006303