A12699.14
A12699.22
A12699.36
A12699.14EA
50.1
EUR
InStock
A12699.14
A12699.22
A12699.36
Amantadin Hydrochlorid 99%
Amantadin Hydrochlorid
Purity: 99%
CAS No.: 665-66-7
Molecular Formula: C10H17N.HCl
Formula Weight: 187.71
Melting Point: 360°
Beilstein Reference: 4198854
Merck Reference: 14,374
MDL No.: MFCD00074723