A12757.30
A12757.0B
A12757.0I
A12757.30EA
35
EUR
InStock
A12757.30
A12757.0B
A12757.0I
4-Chlorbenzaldehyd ≥98%
4-Chlorbenzaldehyd
Purity: 98%
CAS No.: 104-88-1
Molecular Formula: C7H5ClO
Formula Weight: 140.57
Flash Point: 88°(190°F)
Odour: Pungent
Boiling Point: 213-214°
Beilstein Reference: 385858
UN No.: 3077
MDL No.: MFCD00003379