A12787.14
A12787.14EA
322
EUR
InStock
A12787.14
Difluoressigsäure ≥98%
Difluoressigsäure
Purity: 98%
CAS No.: 381-73-7
Molecular Formula: F2CHCO2H
Formula Weight: 96.03
Flash Point: 78°(172°F)
Refractive Index: 1.3430
Odour: Pungent
Melting Point: -1°
Boiling Point: 132-134°
Beilstein Reference: 1098588
UN No.: 3265
MDL No.: MFCD00004220