A12993.03
A12993.06
A12993.14
A12993.03EA
84
EUR
InStock
A12993.03
A12993.06
A12993.14
2,3-Diaminonaphthalin 97%
2,3-Diaminonaphthalin
Purity: 97%
CAS No.: 771-97-1
Molecular Formula: C10H10N2
Formula Weight: 158.20
Melting Point: 193-196°
Solubility: Slightly soluble in water
Beilstein Reference: 2206394
MDL No.: MFCD00004116