A13027.09
A13027.18
A13027.30
A13027.09EA
42.2
EUR
InStock
A13027.09
A13027.18
A13027.30
3,4-Dimethylphenylhydraziniumchlorid ≥98%
3,4-Dimethylphenylhydraziniumchlorid
Purity: 98%
CAS No.: 60481-51-8
Molecular Formula: C8H12N2.HCl
Formula Weight: 172.66
Melting Point: ca 195° dec.
Beilstein Reference: 6096060