A13057.0E
A13057.22
A13057.36
A13057.0EEA
134
EUR
InStock
A13057.0E
A13057.22
A13057.36
Ethylbenzoat ≥99%
Ethylbenzoat
Purity: 99%
CAS No.: 93-89-0
Molecular Formula: C9H10O2
Formula Weight: 150.18
Flash Point: 84°(183°F)
Refractive Index: 1.5050
Melting Point: -34°
Boiling Point: 212-214°
Beilstein Reference: 1908172
Merck Reference: 14,3766
MDL No.: MFCD00009109