A13176.09
A13176.18
A13176.09EA
31.2
EUR
InStock
A13176.09
A13176.18
(2-Chlorphenyl)acetonitril ≥98%
(2-Chlorphenyl)acetonitril
Purity: 98+%
CAS No.: 2856-63-5
Molecular Formula: C8H6ClN
Formula Weight: 151.60
Flash Point: 110°(230°F)
Refractive Index: 1.5440
Melting Point: 21-24°
Boiling Point: 240-242°
Beilstein Reference: 2043683
UN No.: 3439
MDL No.: MFCD00001898