A13192.06
A13192.14
A13192.22
A13192.06EA
40.8
EUR
InStock
A13192.06
A13192.14
A13192.22
1,3-Dichlor-5-iodbenzol ≥99%
1,3-Dichlor-5-iodbenzol
Purity: 99%
CAS No.: 3032-81-3
Molecular Formula: C6H3Cl2I
Formula Weight: 272.90
Melting Point: 55-59°
Boiling Point: 127°/20mm
MDL No.: MFCD00001047