A13255.03
A13255.06
A13255.03EA
24.8
EUR
InStock
A13255.03
A13255.06
2-(4-Fluorphenyl)indol ≥99%
2-(4-Fluorophenyl)indole
Purity: 99%
CAS No.: 782-17-2
Molecular Formula: C14H10FN
Formula Weight: 211.24
Odour: Odourless
Melting Point: 186-191°
Beilstein Reference: 744766
MDL No.: MFCD00068178