A13396.14
A13396.22
A13396.36
AB109247.0100
A13396.14EA
37.4
EUR
InStock
A13396.14
A13396.22
A13396.36
ABCRAB109247.0100
3,4-Dimethoxybenzylalkohol ≥97%
3,4-Dimethoxybenzylalkohol
Purity: 97%
CAS No.: 93-03-8
Molecular Formula: C9H12O3
Formula Weight: 168.19
Flash Point: 110°(230°F)
Refractive Index: 1.5530
Melting Point: ca 22°
Boiling Point: 297-298°
Beilstein Reference: 639388
MDL No.: MFCD00004638