A13421.18
A13421.30
A13421.18EA
44.3
EUR
InStock
A13421.18
A13421.30
Cycloheptanon ≥98%
Cycloheptanon
Purity: 99%
CAS No.: 502-42-1
Molecular Formula: C7H12O
Formula Weight: 112.17
Flash Point: 55°(131°F)
Refractive Index: 1.4610
Odour: Sweet
Melting Point: -21°
Boiling Point: 179-181°
Beilstein Reference: 969823
Merck Reference: 14,2722
UN No.: 1224
MDL No.: MFCD00004159