A13426.22
A13426.22EA
0
EUR
InStock
A13426.22
1-Benzyl-3-pyrrolidinon ≥98%
1-Benzyl-3-pyrrolidinon
Purity: 98%
CAS No.: 775-16-6
Molecular Formula: C11H13NO
Formula Weight: 175.23
Storage Temperature: 2°C to 8°C
Flash Point: 110°(230°F)
Boiling Point: 77-78°/0.01mm
MDL No.: MFCD00005342