A13580.14
A13580.22
A13580.36
A13580.14EA
21.5
EUR
InStock
A13580.14
A13580.22
A13580.36
4-Hydroxybenzaldehyd ≥98%
4-Hydroxybenzaldehyd
Purity: 98%
CAS No.: 123-08-0
Molecular Formula: C7H6O2
Formula Weight: 122.12
Flash Point: 174°(345°F)
Melting Point: 115-119°
Boiling Point: 191°/50mm
Beilstein Reference: 471352
Merck Reference: 14,4811
MDL No.: MFCD00006939