A13614.0E
A13614.14
A13614.22
A13614.36
A13614.0EEA
767
EUR
InStock
A13614.0E
A13614.14
A13614.22
A13614.36
Trifluoressigsäureanhydrid ≥99%
Trifluoressigsäureanhydrid
Purity: 99+%
CAS No.: 407-25-0
Molecular Formula: C4F6O3
Formula Weight: 210.03
Storage Temperature: 2°C to 8°C
Refractive Index: 1.2690
Melting Point: -65°
Boiling Point: 39-40°
Beilstein Reference: 746197
UN No.: 3265
MDL No.: MFCD00000416