A13645.14
A13645.06
A13645.14EA
171
EUR
InStock
A13645.14
A13645.06
2-Brombenzolsulfonylchlorid ≥98%
2-Brombenzolsulfonylchlorid
Purity: 98%
CAS No.: 2905-25-1
Molecular Formula: C6H4BrClO2S
Formula Weight: 255.52
Melting Point: 48-52°
Boiling Point: 127-128°/2.4mm
UN No.: 3261
MDL No.: MFCD00051971