A13675.14
A13675.22
A13675.36
A13675.14EA
30.9
EUR
InStock
A13675.14
A13675.22
A13675.36
2-Methoxypyridin ≥98%
2-Methoxypyridine
Purity: 98%
CAS No.: 1628-89-3
Molecular Formula: C6H7NO
Formula Weight: 109.13
Flash Point: 32°(89°F)
Refractive Index: 1.5040
Boiling Point: 142-143°
Beilstein Reference: 108189
UN No.: 3271
MDL No.: MFCD00006262