A13691.06
A13691.14
A13691.22
A13691.06EA
29.1
EUR
InStock
A13691.06
A13691.14
A13691.22
2-Chlor-N,N-diethylacetamid ≥97%
2-Chlor-N,N-diethylacetamid
Purity: 97%
CAS No.: 2315-36-8
Molecular Formula: C6H12ClNO
Formula Weight: 149.62
Flash Point: 113°(235°F)
Refractive Index: 1.4700
Boiling Point: 123-124°/15mm
Beilstein Reference: 605866
UN No.: 2810
MDL No.: MFCD00000928