A13739.03
A13739.06
A13739.03EA
66.8
EUR
InStock
A13739.03
A13739.06
5-Isoxazolcarbonsäure ≥98%
5-Isoxazolecarboxylic acid
Purity: 98%
CAS No.: 21169-71-1
Molecular Formula: C4H3NO3
Formula Weight: 113.07
Melting Point: 141-144°
MDL No.: MFCD00156151