A13741.06
A13741.14
A13741.06EA
21.4
EUR
InStock
A13741.06
A13741.14
5-Acetyl-2,4-dimethylthiazol ≥99%
5-Acetyl-2,4-dimethylthiazol
Purity: 99%
CAS No.: 38205-60-6
Molecular Formula: C7H9NOS
Formula Weight: 155.22
Flash Point: 104°(219°F)
Refractive Index: 1.5430
Boiling Point: 230°
Beilstein Reference: 120560
MDL No.: MFCD00005331