A13759.06
A13759.14
A13759.06EA
62.4
EUR
InStock
A13759.06
A13759.14
2-Isobutylthiazol ≥99%
2-Isobutylthiazol
Purity: 99%
CAS No.: 18640-74-9
Molecular Formula: C7H11NS
Formula Weight: 141.24
Flash Point: 58°(136°F)
Refractive Index: 1.4960
Odour: Characteristic
Boiling Point: 172-173°
Beilstein Reference: 507823
UN No.: 1993
MDL No.: MFCD00005334