A13776.0B
A13776.18
A13776.22
A13776.30
A13776.36
A13776.0BEA
198
EUR
InStock
A13776.0B
A13776.18
A13776.22
A13776.30
A13776.36
Diethylethoxymethylenmalonat ≥98%
Diethylethoxymethylenmalonat
Purity: 98+%
CAS No.: 87-13-8
Molecular Formula: C10H16O5
Formula Weight: 216.23
Flash Point: 144°(291°F)
Refractive Index: 1.4620
Odour: Ester-like
Melting Point: -33°
Boiling Point: 136-139°/10mm
Beilstein Reference: 880058
MDL No.: MFCD00009148