A13982.06
A13982.14
A13982.06EA
24.1
EUR
InStock
A13982.06
A13982.14
Pyrazin, (max. 1% H₂O) 99+% (Trockengewicht)
Pyrazine
Purity: 99+%
CAS No.: 290-37-9
Molecular Formula: C4H4N2
Formula Weight: 80.09
Flash Point: 55°(131°F)
Refractive Index: 1.5235
Odour: Strong
Melting Point: 50-56°
Boiling Point: 113-116°
Beilstein Reference: 103905
Merck Reference: 14,7957
UN No.: 1325
MDL No.: MFCD00006122