A14003.22
A14003.36
A14003.22EA
44.9
EUR
InStock
A14003.22
A14003.36
1,4-Diazabicyclo[2.2.2]octan ≥98%
1,4-Diazabicyclo[2.2.2]octan
Purity: 98%
CAS No.: 280-57-9
Molecular Formula: C6H12N2
Formula Weight: 112.17
Flash Point: 62°(143°F)
Odour: Amine-like
Boiling Point: 174-176°
Beilstein Reference: 103618
Merck Reference: 14,9669
UN No.: 1325
MDL No.: MFCD00006689