A14005.36
A14005.0E
A14005.18
A14005.30
A14005.36EA
50.6
EUR
InStock
A14005.36
A14005.0E
A14005.18
A14005.30
Hydrazin-Hydrat 98+%
Hydrazin-Hydrat
Purity: 98+%
CAS No.: 7803-57-8
Molecular Formula: H4N2.H2O
Formula Weight: 50.06 (32.05anhy)
Flash Point: 75°(167°F)
Refractive Index: 1.4310
Odour: Amine-like
Melting Point: -52°
Boiling Point: 120-121°
Merck Reference: 14,4771
UN No.: 2030
MDL No.: MFCD00149931