A14122.30
A14122.0B
A14122.18
A14122.30EA
63.1
EUR
InStock
A14122.30
A14122.0B
A14122.18
1-Bromnaphthalin ≥97%
1-Bromnaphthalin
Purity: 97%
CAS No.: 90-11-9
Molecular Formula: C10H7Br
Formula Weight: 207.08
Flash Point: 110°(230°F)
Physical Form: Liquid
Density: 1.485
Refractive Index: 1.6570
Appearance: Colourless to pale yellow
Melting Point: -2° to 0°
Boiling Point: 279-281°
Beilstein Reference: 1906414
Merck Reference: 14,1425
MDL No.: MFCD00003868