A14309.03
A14309.06
A14309.03EA
57.1
EUR
InStock
A14309.03
A14309.06
5,6-Dichloro-2-mercaptobenzimidazole 98%
5,6-Dichloro-2-mercaptobenzimidazole
Purity: 98%
CAS No.: 19462-98-7
Molecular Formula: C7H4Cl2N2S
Formula Weight: 219.09
Odour: Odourless
Melting Point: ca 350° dec.
MDL No.: MFCD00052398