A14319.14
A14319.06
A14319.14EA
225
EUR
InStock
A14319.14
A14319.06
4-Brom-2-methyl-6-nitroanilin ≥97%
4-Brom-2-methyl-6-nitroanilin
Purity: 97%
CAS No.: 77811-44-0
Molecular Formula: C7H7BrN2O2
Formula Weight: 231.06
Melting Point: 143-147°
Beilstein Reference: 3530524