A14331.03
A14331.06
A14331.14
A14331.03EA
26.5
EUR
InStock
A14331.03
A14331.06
A14331.14
5,5'-Dithiobis(2-nitrobenzoesäure) (Ellman's Reagenz, DTNB) ≥99%
5,5'-Dithiobis(2-nitrobenzoesäure) (Ellman's Reagenz, DTNB)
Purity: 99%
CAS No.: 69-78-3
Molecular Formula: C14H8N2O8S2
Formula Weight: 396.36
Melting Point: ca 240° dec.
Beilstein Reference: 1896821