A14351.09
A14351.18
A14351.30
A14351.09EA
39.1
EUR
InStock
A14351.09
A14351.18
A14351.30
4-Isopropoxybenzoesäure ≥99%
4-Isopropoxybenzoesäure
Purity: 99%
CAS No.: 13205-46-4
Molecular Formula: C10H12O3
Formula Weight: 180.20
Odour: Odourless
Melting Point: 163-167°
MDL No.: MFCD00044318