A14413.09
A14413.18
A14413.30
A14413.09EA
40.9
EUR
InStock
A14413.09
A14413.18
A14413.30
2-Nitro-4-(trifluormethyl)anilin ≥98%
2-Nitro-4-(trifluormethyl)anilin
Purity: 98%
CAS No.: 400-98-6
Molecular Formula: C7H5F3N2O2
Formula Weight: 206.13
Melting Point: 105-109°
Beilstein Reference: 650808
UN No.: 2811
MDL No.: MFCD00007155