A14417.06
A14417.03
A14417.06EA
247
EUR
InStock
A14417.06
A14417.03
2-(Phosphonooxy)benzoesäure 98%
2-(Phosphonooxy)benzoic acid
Purity: 98%
CAS No.: 6064-83-1
Molecular Formula: C7H7O6P
Formula Weight: 218.10
Melting Point: 162-166°
Beilstein Reference: 2619142
MDL No.: MFCD00042645