Stabilization:
Stabilisiert mit Diethylether
A14420.14
A14420.22
A14420.14EA
79.9
EUR
InStock
A14420.14
A14420.22
Triethyloxoniumtetrafluoroborat ≥95% stabilisiert
Triethyloxonium tetrafluoroborate
Stabilized with 3-5% diethyl ether
Purity: 95%
CAS No.: 368-39-8
Molecular Formula: C6H15BF4O
Formula Weight: 189.99
Storage Temperature: 2°C to 8°C
Melting Point: 84-89°
Beilstein Reference: 3598090
UN No.: 2925
MDL No.: MFCD00044423