A14425.14
A14425.22
A14425.36
A14425.14EA
51.6
EUR
InStock
A14425.14
A14425.22
A14425.36
Methyl-3-hydroxybenzoat ≥99%
Methyl-3-hydroxybenzoat
Purity: 99%
CAS No.: 19438-10-9
Molecular Formula: C8H8O3
Formula Weight: 152.15
Melting Point: 70-72°
Boiling Point: 280-281°/708mm
MDL No.: MFCD00002295