A14677.14
A14677.06
A14677.14EA
310
EUR
InStock
A14677.14
A14677.06
5-Brom-2-thiophensulfonylchlorid ≥98%
5-Brom-2-thiophensulfonylchlorid
Purity: 98%
CAS No.: 55854-46-1
Molecular Formula: C4H2BrClO2S2
Formula Weight: 261.55
Melting Point: 39-42°
Boiling Point: 99-101°/0.5mm
UN No.: 3261
MDL No.: MFCD00084909