Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Iodacetamid 98% stabilisiert

Lieferant: ALFA AESAR

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

Gefahr

Stabilization: ∼5-8% H2O

A14715.06 A14715.14 A14715.22
A14715.06EA 25.6 EUR InStock
A14715.06 A14715.14 A14715.22
Iodacetamid 98% stabilisiert
Iodacetamid
Alkylating agent used in peptide mapping because it covalently binds with the thiols in cysteine to prevent disulfide bond formation.

2-Iodoacetamide is used as an electrophile for covalent modification of nucleophilic residues on proteins such as cysteine, methionine and histidine. It is used to bind with thiol group of cysteine, thereby it protects the formation disulfide bonds. It is involved as an inhibitor of deubiquitinase enzymes (DUBs). Further, it acts as an alkylating sulfhydryl reagent.

Alkylating agent used in peptide mapping because it covalently binds with the thiols in cysteine to prevent disulfide bond formation


Formel: ICH₂CONH₂
MW: 184.96 g/mol
Siedepunkt: 266
Melting Pt: 92…95 °C (getrocknet)
Storage Temperature: Kühlschrank
MDL Number: MFCD00008028
CAS-Nummer: 144-48-9
EINECS: 205-630-1
UN: 2811
ADR: 6.1 III
Order Now

 

PRODUKT

TECHNISCHE DATEN

WEITERE INFORMATIONEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge