A14719.0E
A14719.22
A14719.36
A14719.0EEA
388
EUR
InStock
A14719.0E
A14719.22
A14719.36
6-Aminocapronsäure 99%
6-Aminocapronsäure
Purity: 99%
CAS No.: 60-32-2
Molecular Formula: C6H13NO2
Formula Weight: 131.18
Physical Form: Cryst powder
Appearance: White
Melting Point: 210-212°
Beilstein Reference: 906872
Merck Reference: 14,432
MDL No.: MFCD00008238