A14728.0C
A14728.0E
A14728.30
A14728.36
AB115885.0250
A14728.0CEA
209
EUR
InStock
A14728.0C
A14728.0E
A14728.30
A14728.36
ABCRAB115885.0250
Diethylenglycol ≥99%
Diethylenglycol
Purity: 99%
CAS No.: 111-46-6
Molecular Formula: C4H10O3
Formula Weight: 106.12
Flash Point: 143°(289°F)
Physical Form: Clear liquid
Density: 1.118
Refractive Index: 1.4470
Sensitivity: Hygroscopic
Appearance: Colourless
Odour: Nearly Odourless
Melting Point: -10°
Boiling Point: 245-246°
Beilstein Reference: 969209
Merck Reference: 14,3119
MDL No.: MFCD00002882