A14768.06
A14768.14
A14768.06EA
34.5
EUR
InStock
A14768.06
A14768.14
(3-Pyridyl)essigsäureethylester ≥99%
(3-Pyridyl)essigsäureethylester
Purity: 99%
CAS No.: 39931-77-6
Molecular Formula: C9H11NO2
Formula Weight: 165.19
Flash Point: 110°(230°F)
Refractive Index: 1.500
Boiling Point: 79-80°/0.4mm
Beilstein Reference: 123827
MDL No.: MFCD00006411