A14780.09
A14780.18
A14780.30
AB132028.0250
A14780.09EA
42.5
EUR
InStock
A14780.09
A14780.18
A14780.30
ABCRAB132028.0250
Monomethylphthalat ≥97%
Monomethylphthalat
Purity: 97%
CAS No.: 4376-18-5
Molecular Formula: C9H8O4
Formula Weight: 180.16
Melting Point: 81-84°
Beilstein Reference: 2048766