A14843.0B
A14843.14
A14843.22
A14843.0BEA
778
EUR
InStock
A14843.0B
A14843.14
A14843.22
4-(Benzyloxy)phenol ≥98%
4-Benzyloxyphenol
Purity: 98+%
CAS No.: 103-16-2
Molecular Formula: C13H12O2
Formula Weight: 200.24
Melting Point: 118-122°
Beilstein Reference: 1958305
Merck Reference: 14,6248
MDL No.: MFCD00002333