A14870.09
A14870.18
A14870.30
A14870.09EA
43.7
EUR
InStock
A14870.09
A14870.18
A14870.30
2,3,5-Trimethylpyrazin ≥99%
2,3,5-Trimethylpyrazine
Purity: 99%
CAS No.: 14667-55-1
Molecular Formula: C7H10N2
Formula Weight: 122.17
Flash Point: 54°(129°F)
Refractive Index: 1.5050
Boiling Point: 171-172°
Beilstein Reference: 2423
UN No.: 1993
MDL No.: MFCD00006145